Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

1-Hexyl-4-iodobenzene 98%

Supplier: Thermo Scientific Chemicals
Warning

H26878-03 H26878-09
AAH26878-03EA 42.99 USD InStock
AAH26878-03 AAH26878-09
1-Hexyl-4-iodobenzene 98%
1-Hexyl-4-iodobenzene


Formula: C₁₂H₁₇I
MW: 288.17 g/mol
Boiling Pt: 122…125 °C/3 mm
Flash Pt: >120 °C(248 °F)
Storage Temperature: Ambient
MDL Number: MFCD07777097
CAS Number: 62150-34-9
Order Now

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty