Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

D-Valine methyl ester hydrochloride 99%

Supplier: Thermo Scientific Chemicals
Synonyms: H-D-Val-OMe.HCl

H26883-06 H26883-14 H26883-03
AAH26883-06EA 125.99 USD InStock
AAH26883-06 AAH26883-14 AAH26883-03
D-Valine methyl ester hydrochloride 99%
D-Valine methyl ester hydrochloride


Formula: C₆H₁₃NO₂
MW: 167.633 g/mol
Melting Pt: ca 170 °C (dec.)
Storage Temperature: Ambient
MDL Number: MFCD00237309
CAS Number: 7146-15-8
EINECS: 230-454-7
Order Now

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty