Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

1-Butylpyridinium bromide 99%

Supplier: Thermo Scientific Chemicals
Warning
Synonyms: 1-n-Butylpyridinium bromideN-butylpyridinium bromide

H26995-18 H26995-06
AAH26995-18EA 200.99 USD InStock
AAH26995-18 AAH26995-06
1-Butylpyridinium bromide 99%
1-Butylpyridinium bromide


Formula: C₉H₁₄BrN
MW: 216.12 g/mol
Melting Pt: 105…108 °C
Storage Temperature: Ambient
MDL Number: MFCD00671496
CAS Number: 874-80-6
Order Now

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty