Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

D-Cysteine hydrochloride monohydrate 99%

Supplier: Thermo Scientific Chemicals
Warning
Synonyms: H-D-Cys-OH.HCl(S)-2-Amino-3-mercaptopropanoic acid hydrochloride hydrate

H27107-03 H27107-06
AAH27107-03EA 102.99 USD InStock
AAH27107-03 AAH27107-06
D-Cysteine hydrochloride monohydrate 99%
D-Cysteine hydrochloride monohydrate


Formula: C₃H₇NO₂S·HCl·H₂O
MW: 175.64 g/mol (157.62 anhy)
Melting Pt: 184…186 °C
Storage Temperature: Ambient
MDL Number: MFCD00150051
CAS Number: 207121-46-8
EINECS: 251-043-9
Order Now

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty