Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

6-Acetyl-2(3H)-benzothiazolone 97%

Supplier: Thermo Scientific Chemicals
Warning

H27154-03 H27154-09
AAH27154-03EA 32.79 USD InStock
AAH27154-03 AAH27154-09
6-Acetyl-2(3H)-benzothiazolone 97%
6-Acetyl-2(3H)-benzothiazolone


Formula: C₉H₇NO₂S
MW: 193.22 g/mol
Melting Pt: 190…194 °C
Storage Temperature: Ambient
MDL Number: MFCD02660572
CAS Number: 133044-44-7
Order Now

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty