Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

6-Nitrochromone 97%

Supplier: Thermo Scientific Chemicals
Warning
Synonyms: 6-Nitro-4H-chromen-4-one

H27167-03 H27167-06 H27167-14
AAH27167-06EA 74.79 USD InStock
AAH27167-06 AAH27167-03 AAH27167-14
6-Nitrochromone 97%
6-Nitrochromone


Formula: C₉H₅NO₄
MW: 191.15 g/mol
Melting Pt: 172…175 °C
Storage Temperature: Ambient
MDL Number: MFCD02954226
CAS Number: 51484-05-0
Order Now

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty