Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

3-Bromomethylquinoxaline-2-one 90%, tech.

Supplier: Thermo Scientific Chemicals
Danger

H27169-06 H27169-14
AAH27169-14EA 104.99 USD InStock
AAH27169-14 AAH27169-06
3-Bromomethylquinoxaline-2-one 90%, tech.
3-Bromomethylquinoxaline-2-one


Formula: C₉H₇BrN₂O
MW: 239.07 g/mol
Melting Pt: >300 °C (dec.)
Storage Temperature: Ambient
MDL Number: MFCD03412060
CAS Number: 62235-61-4
UN: 1759
ADR: "8",III
Order Now

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty