Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

1-Butyl-1-methylpyrrolidinium bis(trifluoromethylsulfonyl)imide 98%

Supplier: Thermo Scientific Chemicals
Warning
Synonyms: N-Butyl-N-methylpyrrolidinium bis((trifluoromethyl)sulphonyl)imideN-Butyl-N-methylpyrrolidinium bis((trifluoromethyl)sulfonyl)imide

H27177-06 H27177-18
AAH27177-18EA 496.99 USD InStock
AAH27177-18 AAH27177-06
1-Butyl-1-methylpyrrolidinium bis(trifluoromethylsulfonyl)imide 98%
1-Butyl-1-methylpyrrolidinium bis(trifluoromethylsulphonyl)imide


Formula: C₁₁H₂₀F₆N₂O₄S₂
MW: 422.41 g/mol
Storage Temperature: Ambient
MDL Number: MFCD07784447
CAS Number: 223437-11-4
Order Now

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty