Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

1-O-Methyl-2-deoxy-D-ribose 96%

Supplier: Thermo Scientific Chemicals
Synonyms: (2R,3S)-2-(Hydroxymethyl)-5-methoxytetrahydrofuran-3-ol

H27181-06 H27181-03
AAH27181-03EA 80.99 USD InStock
AAH27181-03 AAH27181-06
1-O-Methyl-2-deoxy-D-ribose 96%
1-O-Methyl-2-deoxy-D-ribose


Formula: C₆H₁₂O₄
MW: 148.17 g/mol
Density: 1.224
Flash Pt: >110 °C(230 °F)
Storage Temperature: Ambient
MDL Number: MFCD00134161
CAS Number: 60134-26-1
Order Now

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty