Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

L-m-Tyrosine ≥97%

Supplier: Thermo Scientific Chemicals

H27224-MD H27224-03
AAH27224-03EA 414.99 USD InStock
AAH27224-03 AAH27224-MD
L-m-Tyrosine ≥97%
L-m-Tyrosine


Formula: C₉H₁₁NO₃
MW: 181.19 g/mol
Melting Pt: 260…270 °C
Storage Temperature: Ambient
MDL Number: MFCD00063059
CAS Number: 587-33-7
Order Now

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty