Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

4-Chloro-o-toluic acid 98%

Supplier: Thermo Scientific Chemicals
Danger

H27262-06 H27262-03
AAH27262-03EA 97.09 USD InStock
AAH27262-03 AAH27262-06
4-Chloro-o-toluic acid 98%
4-Chloro-o-toluic acid


Formula: C₈H₇ClO₂
MW: 170.6 g/mol
Melting Pt: 170…174 °C
Storage Temperature: Ambient
MDL Number: MFCD00045852
CAS Number: 7499-07-2
Order Now

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty