Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

1-Aminocyclopropanecarboxylic acid hydrochloride 97%

Supplier: Thermo Scientific Chemicals
Warning
Synonyms: 1-azanylcyclopropane-1-carboxylic acid hydrochloride1-aminocyclopropane-1-carboxylic acid hydrochlorideACPCCyclopropanecarboxylic acid, 1-amino-, hydrochloride

H27264-MD H27264-03
AAH27264-03EA 405.99 USD InStock
AAH27264-03 AAH27264-MD
1-Aminocyclopropanecarboxylic acid hydrochloride 97%
1-Aminocyclopropanecarboxylic acid hydrochloride


Formula: C₄H₈ClNO₂
MW: 137.57 g/mol
Melting Pt: ca 220 °C (dec.)
Storage Temperature: Ambient
MDL Number: MFCD00012545
CAS Number: 68781-13-5
Order Now

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty