Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

2-Amino-5-bromopyrimidine 97%

Supplier: Thermo Scientific Chemicals
Warning
Synonyms: 5-Bromopyrimidin-2-ylamine5-Bromopyrimidin-2-amine

H27291-06 H27291-03
AAH27291-06EA 142.99 USD InStock
AAH27291-06 AAH27291-03
2-Amino-5-bromopyrimidine 97%
2-Amino-5-bromopyrimidine


Formula: C₄H₄BrN₃
MW: 174 g/mol
Melting Pt: 232…234 °C
Storage Temperature: Ambient
MDL Number: MFCD00012341
CAS Number: 7752-82-1
Order Now

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty