Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

(R)-1-(Boc-amino)-2-propanol 98%

Supplier: Thermo Scientific Chemicals
Synonyms: (R)-tert-Butyl (2-hydroxypropyl)carbamate

H27296-03 H27296-06
AAH27296-06EA 334.99 USD InStock
AAH27296-06 AAH27296-03
(R)-1-(Boc-amino)-2-propanol 98%
(R)-1-(Boc-amino)-2-propanol


Formula: C₈H₁₇NO₃
MW: 175.23 g/mol
Storage Temperature: Ambient
MDL Number: MFCD04974338
CAS Number: 119768-44-4
Order Now
Specification Test Results
Optical rotation -24° (c=1 in chloroform)

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty