Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

3-Maleimidopropionic acid N-hydroxysuccinimide ester 99%

Supplier: Thermo Scientific Chemicals
Warning
Synonyms: BMPSN-(3-Maleimidopropionyloxy)succinimideN-Succinimidyl 3-maleimidopropionate3-Maleimidopropionic Acid N-Succinimidyl EsterN-β-maleimidopropyl-oxysuccinimide esterN-Succinimidyl 3-Maleimidopropionat

H27329-MD H27329-03
AAH27329-03EA 566.11 USD InStock
AAH27329-03 AAH27329-MD
3-Maleimidopropionic acid N-hydroxysuccinimide ester 99%
BMPS (3-Maleimidopropionic acid N-hydroxysuccinimide ester)


Formula: C₁₁H₁₀N₂O₆
MW: 266.21 g/mol
Melting Pt: 168…170 °C
Storage Temperature: Ambient
MDL Number: MFCD00043141
CAS Number: 55750-62-4
Order Now

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty