Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

1-Pentyl-4-iodobenzene 98%

Supplier: Thermo Scientific Chemicals
Warning
Synonyms: 1-Iodo-4-pentylbenzene4-Pentyliodbenzol1-Iodo-4-n-pentylbenzene4-Iodopentylbenzene4-Pentyliodobenzene1-(4-Iodophenyl)pentane

H27356-03 H27356-09
AAH27356-03EA 37.69 USD InStock
AAH27356-03 AAH27356-09
1-Pentyl-4-iodobenzene 98%
1-Pentyl-4-iodobenzene


Formula: C₁₁H₁₅I
MW: 274.14 g/mol
Boiling Pt: 88 °C/1 mm
Density: 1.38
Flash Pt: >110 °C(230 °F)
Storage Temperature: Ambient
MDL Number: MFCD00079767
CAS Number: 85017-60-3
Order Now

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty