Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

4-Amino-o-anisic acid 97%

Supplier: Thermo Scientific Chemicals
Warning

H27419-06 H27419-14
AAH27419-14EA 74.19 USD InStock
AAH27419-14 AAH27419-06
4-Amino-o-anisic acid 97%
4-Amino-o-anisic acid


Formula: C₈H₉NO₃
MW: 167.16 g/mol
Melting Pt: ca 150 °C (dec.)
Storage Temperature: Ambient
MDL Number: MFCD00114479
CAS Number: 2486-80-8
EINECS: 219-632-5
Order Now

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty