Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

N-Acetyl-DL-leucine 99%

Supplier: Thermo Scientific Chemicals
Synonyms: 2-Acetamido-4-methylpentanoic acidAc-DL-Leu-OH

H27441-14 H27441-22 H27441-06
AAH27441-06EA 52.49 USD InStock
AAH27441-06 AAH27441-14 AAH27441-22
N-Acetyl-DL-leucine 99%
N-Acetyl-DL-leucine


Formula: C₈H₁₅NO₃
MW: 173.2 g/mol
Melting Pt: 159…161 °C
Storage Temperature: Ambient
MDL Number: MFCD00026498
CAS Number: 99-15-0
EINECS: 202-734-9
Order Now

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty