Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

3-(Methoxycarbonyl)phenylboronic acid 97%

Supplier: Thermo Scientific Chemicals
Warning
Synonyms: 3-(Methoxycarbonyl)benzeneboronic acid

H27444-06 H27444-03
AAH27444-03EA 58.89 USD InStock
AAH27444-03 AAH27444-06
3-(Methoxycarbonyl)phenylboronic acid 97%
3-(Methoxycarbonyl)phenylboronic acid


Formula: C₈H₉BO₄
MW: 179.97 g/mol
Melting Pt: 136 °C
Storage Temperature: Ambient
MDL Number: MFCD02093046
CAS Number: 99769-19-4
Order Now

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty