Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

2-Amino-3-nitrophenol 98%

Supplier: Thermo Scientific Chemicals
Warning
Synonyms: 2-Hydroxy-6-nitroaniline

H27472-14 H27472-06
AAH27472-06EA 75.99 USD InStock
AAH27472-06 AAH27472-14
2-Amino-3-nitrophenol 98%
2-Amino-3-nitrophenol


Formula: C₆H₆N₂O₃
MW: 154.13 g/mol
Melting Pt: ca 212 °C
Storage Temperature: Ambient
MDL Number: MFCD00010875
CAS Number: 603-85-0
EINECS: 210-060-1
Order Now

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty