Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

6-Bromo-2H-1,4-benzoxazin-3(4H)-one 95%

Supplier: Thermo Scientific Chemicals
Warning

H27493-03 H27493-09
AAH27493-03EA 58.39 USD InStock
AAH27493-03 AAH27493-09
6-Bromo-2H-1,4-benzoxazin-3(4H)-one 95%
6-Bromo-2H-1,4-benzoxazin-3(4H)-one


Formula: C₈H₆BrNO₂
MW: 228.04 g/mol
Melting Pt: 220…225 °C
Storage Temperature: Ambient
MDL Number: MFCD00461173
CAS Number: 24036-52-0
Order Now

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty