Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

5-Methoxy-m-cresol 97%

Supplier: Thermo Scientific Chemicals
Warning
Synonyms: 3-Hydroxy-5-methylanisol1-Hydroxy-3-methoxy-5-methylbenzene3-Hydroxy-5-methoxytoluene3-Methoxy-5-methylphenol

H27516-03 H27516-06
AAH27516-06EA 169.99 USD InStock
AAH27516-06 AAH27516-03
5-Methoxy-m-cresol 97%
5-Methoxy-m-cresol


Formula: C₈H₁₀O₂
MW: 138.17 g/mol
Melting Pt: 60…66 °C
Storage Temperature: Ambient
MDL Number: MFCD00059261
CAS Number: 3209-13-0
EINECS: 221-716-1
Order Now

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty