Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

3-Amino-2-methylpyridine 97%

Supplier: Thermo Scientific Chemicals
Warning
Synonyms: 3-Amino-2-picoline 2-Methyl-3-pyridylamine 2-Methylpyridin-3-amine

H27540-MD H27540-03
AAH27540-03EA 205.99 USD InStock
AAH27540-03 AAH27540-MD
3-Amino-2-methylpyridine 97%
3-Amino-2-methylpyridine


Formula: C₆H₈N₂
MW: 108.14 g/mol
Melting Pt: 114…119 °C
Storage Temperature: Ambient
MDL Number: MFCD03788195
CAS Number: 3430-10-2
ADR: 6.1,III
Order Now

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty