Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

4-Bromo-2-nitrobenzoic acid 97%

Supplier: Thermo Scientific Chemicals
Warning

H27541-06 H27541-03
AAH27541-06EA 122.99 USD InStock
AAH27541-06 AAH27541-03
4-Bromo-2-nitrobenzoic acid 97%
4-Bromo-2-nitrobenzoic acid


Formula: C₇H₄BrNO₄
MW: 246.02 g/mol
Melting Pt: 165…169 °C
Storage Temperature: Ambient
MDL Number: MFCD01013599
CAS Number: 99277-71-1
Order Now

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty