Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

4-(2-Aminoethyl)pyridine 97%

Supplier: Thermo Scientific Chemicals
Danger
Synonyms: 2-(4-Pyridyl)ethylamine

H27545-06 H27545-03
AAH27545-06EA 642.99 USD InStock
AAH27545-06 AAH27545-03
4-(2-Aminoethyl)pyridine 97%
4-(2-Aminoethyl)pyridine


Formula: C₇H₁₀N₂
MW: 122.17 g/mol
Boiling Pt: 70…71 °C/1 mm
Melting Pt: 76 °C
Density: 1.021
Flash Pt: >110 °C(230 °F)
Storage Temperature: Ambient
MDL Number: MFCD00038045
CAS Number: 13258-63-4
UN: 2735
ADR: "8",III
Order Now

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty