Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

N-Methylbutan-2-amine hydrochloride 94%

Supplier: Thermo Scientific Chemicals
Warning
Synonyms: 2-(Methylamino)butane hydrochloride2-Methylamino-butane-hydrochloride

H27547-03 H27547-06
AAH27547-06EA 254.99 USD InStock
AAH27547-06 AAH27547-03
N-Methylbutan-2-amine hydrochloride 94%
N-Methylbutan-2-amine hydrochloride


Formula: C₅H₁₃N·HCl
MW: 123.62 g/mol
Melting Pt: 76…82 °C
Storage Temperature: Ambient
MDL Number: MFCD07777098
CAS Number: 75098-40-7
Order Now

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty