Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

N-Methylallylamine 96%

Supplier: Thermo Scientific Chemicals
Danger
Synonyms: N-Allylmethylamine

H27551-03 H27551-06
AAH27551-06EA 225.99 USD InStock
AAH27551-06 AAH27551-03
N-Methylallylamine 96%
N-Methylallylamine


Formula: C₄H₉N
Boiling Pt: 61…62 °C
Density: 0.748
Flash Pt: -8 °C(-18 °F)
Storage Temperature: Ambient
MDL Number: MFCD00008641
CAS Number: 627-37-2
EINECS: 210-996-0
UN: 3286
ADR: "3",II
Order Now

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty