Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

3-Ethyl-5-methylisoxazole-4-carboxylic acid 97%

Supplier: Thermo Scientific Chemicals
Warning

H27574-03 H27574-09
AAH27574-03EA 28.49 USD InStock
AAH27574-03 AAH27574-09
3-Ethyl-5-methylisoxazole-4-carboxylic acid 97%
3-Ethyl-5-methylisoxazole-4-carboxylic acid


Formula: C₇H₉NO₃
Melting Pt: 123…125 °C
Storage Temperature: Ambient
MDL Number: MFCD03422293
CAS Number: 17147-85-2
Order Now

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty