Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

(±)-2,2'-Bis(di-p-tolylphosphino)-1,1'-binaphthyl 98%

Supplier: Thermo Scientific Chemicals
Warning
Synonyms: rac-2,2'-Bis(di-p-tolylphosphino)-1,1'-binaphthyl

H27585-MD H27585-06 H27585-03
AAH27585-06EA 398.99 USD InStock
AAH27585-06 AAH27585-MD AAH27585-03
(±)-2,2'-Bis(di-p-tolylphosphino)-1,1'-binaphthyl 98%
(±)-2,2'-Bis(di-p-tolylphosphino)-1,1'-binaphthyl


Formula: C₄₈H₄₀P₂
MW: 678.79
Melting Pt: 267…271 °C
Storage Temperature: Ambient
MDL Number: MFCD01311709
CAS Number: 153305-67-0
Order Now

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty