Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

2-Chloro-4-fluorophenylboronic acid 98%

Supplier: Thermo Scientific Chemicals
Warning

H27591-06 H27591-03 H27591-14
AAH27591-06EA 259.99 USD InStock
AAH27591-06 AAH27591-03 AAH27591-14
2-Chloro-4-fluorophenylboronic acid 98%
2-Chloro-4-fluorophenylboronic acid


Formula: C₆H₅BClFO₂
MW: 174.37 g/mol
Storage Temperature: Ambient
MDL Number: MFCD02684295
CAS Number: 313545-72-1
Order Now

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty