Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

7-Bromo-3,4-dihydro-2H-1,5-benzodioxepine 96%

Supplier: Thermo Scientific Chemicals
Warning

H27594-03 H27594-06
AAH27594-06EA 129.99 USD InStock
AAH27594-06 AAH27594-03
7-Bromo-3,4-dihydro-2H-1,5-benzodioxepine 96%
7-Bromo-3,4-dihydro-2H-1,5-benzodioxepine


Formula: C₉H₉BrO₂
Boiling Pt: 145…148 °C/16 mm
Flash Pt: >120 °C(248 °F)
Storage Temperature: Ambient
MDL Number: MFCD00508291
CAS Number: 147644-11-9
Order Now

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty