Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

6-Fluoropyridine-2-carbonitrile 95%

Supplier: Thermo Scientific Chemicals
Warning
Synonyms: 6-Fluoropicolinonitrile2-Cyano-6-fluoropyridine

H27600-MD H27600-03 H27600-06
AAH27600-06EA 827.99 USD InStock
AAH27600-06 AAH27600-MD AAH27600-03
6-Fluoropyridine-2-carbonitrile 95%
6-Fluoropyridine-2-carbonitrile


Formula: C₆H₃FN₂
MW: 122.1 g/mol
Melting Pt: 32…35 °C
Storage Temperature: Ambient
MDL Number: MFCD09263323
CAS Number: 3939-15-9
Order Now

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty