Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

D-Homophenylalanine 98%

Supplier: Thermo Scientific Chemicals
Synonyms: (R)-2-Amino-4-phenylbutyric Acid

H27629-06 H27629-03
AAH27629-03EA 116.99 USD InStock
AAH27629-03 AAH27629-06
D-Homophenylalanine 98%
D-Homophenylalanine


Formula: C₁₀H₁₃NO₂
MW: 179.22 g/mol
Melting Pt: >300 °C
Storage Temperature: Ambient
MDL Number: MFCD00063091
CAS Number: 82795-51-5
Order Now

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty