Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

3-Bromopyridine-2-carbonitrile 98%

Supplier: Thermo Scientific Chemicals
Danger
Synonyms: 3-Bromo-2-cyanopyridine

H27632-03 H27632-MD
AAH27632-03EA 115.99 USD InStock
AAH27632-03 AAH27632-MD
3-Bromopyridine-2-carbonitrile 98%
3-Bromopyridine-2-carbonitrile


Formula: C₆H₃BrN₂
MW: 183.01 g/mol
Boiling Pt: 120 °C/4 mm
Melting Pt: 96…99 °C
Storage Temperature: Ambient
MDL Number: MFCD02683288
CAS Number: 55758-02-6
UN: 3439
ADR: "6.1",III
Order Now

 

ORDER

SPECIFICATIONS

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty