Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

cis-4-Hydroxy-D(+)-proline 98+%

Supplier: Thermo Scientific Chemicals
Synonyms: H-D-cis-Hyp-OH(R)-(+)-cis-4-Hydroxyprolinecis-4-Hydroxy-D-prolineD(+)-cis-4-Hydroxyproline

H27633-MD H27633-03
AAH27633-03EA 79.69 USD InStock
AAH27633-03 AAH27633-MD
cis-4-Hydroxy-D(+)-proline 98+%
cis-4-Hydroxy-D(+)-proline


Formula: C₅H₉NO₃
MW: 131.13 g/mol
Melting Pt: 243 °C (dec.)
Storage Temperature: Ambient
MDL Number: MFCD00005252
CAS Number: 2584-71-6
EINECS: 219-963-5
Order Now

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty