Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

1-Ethyl-3-methylimidazolium chloride 98+%

Supplier: Thermo Scientific Chemicals
Warning
Synonyms: 1-Ethyl-3-methyl-1H-imidazol-3-ium chloride

H27651-06 H27651-14
AAH27651-06EA 102.99 USD InStock
AAH27651-06 AAH27651-14
1-Ethyl-3-methylimidazolium chloride 98+%
1-Ethyl-3-methylimidazolium chloride


Formula: C₆H₁₁ClN₂
MW: 146.62 g/mol
Melting Pt: 77…80 °C
Flash Pt: 186 °C(367 °F)
Storage Temperature: Ambient
MDL Number: MFCD00074843
CAS Number: 65039-09-0
Order Now

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty