Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

4-Bromo-2-fluoropyridine 95%

Supplier: Thermo Scientific Chemicals
Warning

H27657-03 H27657-06
AAH27657-03EA 176.49 USD InStock
AAH27657-03 AAH27657-06
4-Bromo-2-fluoropyridine 95%
4-Bromo-2-fluoropyridine


Formula: C₅H₃BrFN
MW: 175.99 g/mol
Storage Temperature: Ambient
MDL Number: MFCD04112504
CAS Number: 128071-98-7
ADR: 6.1,III
Order Now

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty