Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

3-Bromo-2-ethoxypyridine 95%

Supplier: Thermo Scientific Chemicals
Warning

H27690-MD H27690-03
AAH27690-MDEA 94.52 USD InStock
AAH27690-MD AAH27690-03
3-Bromo-2-ethoxypyridine 95%
3-Bromo-2-ethoxypyridine


Formula: C₇H₈BrNO
MW: 202.05 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00234309
CAS Number: 57883-25-7
Order Now

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty