Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

N-Acetyl-DL-serine ≥98%

Supplier: Thermo Scientific Chemicals
Synonyms: Ac-DL-Ser-OH

H27717-06
AAH27717-06EA 401.99 USD InStock
AAH27717-06
N-Acetyl-DL-serine ≥98%
N-Acetyl-DL-serine


Formula: C₅H₉NO₄
MW: 147.1 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD00037248
CAS Number: 97-14-3
Order Now

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty