Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

N-(α)-Boc-D-tryptophanol 98%

Supplier: Thermo Scientific Chemicals
Synonyms: Boc-D-Tryptophanol

H27763-MD H27763-06 H27763-03
AAH27763-03EA 270.99 USD InStock
AAH27763-03 AAH27763-MD AAH27763-06
N-(α)-Boc-D-tryptophanol 98%
N-(alpha)-Boc-D-tryptophanol


Formula: C₁₆H₂₂N₂O₃
MW: 290.3 g/mol
Melting Pt: 119…121 °C
Storage Temperature: Ambient
MDL Number: MFCD00270221
CAS Number: 158932-00-4
Order Now

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty