Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

2-Nitroterephthalic acid-4-methyl ester 97%

Supplier: Thermo Scientific Chemicals
Warning
Synonyms: 4-(Methoxycarbonyl)-2-nitrobenzoic acid

H27779-06 H27779-14
AAH27779-14EA 268.99 USD InStock
AAH27779-14 AAH27779-06
2-Nitroterephthalic acid-4-methyl ester 97%
2-Nitroterephthalic acid 4-methyl ester


Formula: C₉H₇NO₆
MW: 225.17 g/mol
Melting Pt: 132…136 °C
Storage Temperature: Ambient
MDL Number: MFCD06203344
CAS Number: 55737-66-1
Order Now

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty