Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

N-Boc-4-oxo-L-proline methyl ester 97%

Supplier: Thermo Scientific Chemicals
Danger
Synonyms: Boc-4-oxo-Pro-OMe(S)-1-tert-Butyl 2-methyl 4-oxopyrrolidine-1,2-dicarboxylate

H27782-03 H27782-06 H27782-14
AAH27782-06EA 136.99 USD InStock
AAH27782-06 AAH27782-03 AAH27782-14
N-Boc-4-oxo-L-proline methyl ester 97%
N-Boc-4-oxo-L-proline methyl ester


Formula: C₁₁H₁₇NO₅
MW: 243.259 g/mol
Melting Pt: 35…40 °C
Storage Temperature: Refrigerator
MDL Number: MFCD01861778
CAS Number: 102195-80-2
Order Now

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty