Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

6-Nitro-1,3-benzoxazol-2(3H)-one 98%

Supplier: Thermo Scientific Chemicals
Warning
Synonyms: 6-Nitrobenzoxazole-2(3H)-one

H27796-06 H27796-03
AAH27796-06EA 84.29 USD InStock
AAH27796-06 AAH27796-03
6-Nitro-1,3-benzoxazol-2(3H)-one 98%
6-Nitro-1,3-benzoxazol-2(3H)-one


Formula: C₇H₄N₂O₄
MW: 180.12 g/mol
Melting Pt: 244…249 °C
Storage Temperature: Ambient
MDL Number: MFCD00463755
CAS Number: 4694-91-1
EINECS: 225-157-4
Order Now

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty