Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

1-Boc-3-oxopiperazine 98%

Supplier: Thermo Scientific Chemicals
Warning
Synonyms: 4-N-BOC-2-oxo-piperazinetert-butyl 3-oxopiperazine-1-carboxylatetert-butyl 3-oxo-1-piperazinecarboxylate

H27823-03 H27823-06
AAH27823-06EA 290.99 USD InStock
AAH27823-06 AAH27823-03
1-Boc-3-oxopiperazine 98%
1-Boc-3-oxopiperazine


Formula: C₉H₁₆N₂O₃
MW: 200.238 g/mol
Melting Pt: 156…160 °C
Storage Temperature: Ambient
MDL Number: MFCD02181069
CAS Number: 76003-29-7
Order Now

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty