Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

4-Phenyl-3-butyn-2-one 96%

Supplier: Thermo Scientific Chemicals
Warning

H27826-03 H27826-06
AAH27826-03EA 64.31 USD InStock
AAH27826-03 AAH27826-06
4-Phenyl-3-butyn-2-one 96%
4-Phenyl-3-butyn-2-one


Formula: C₁₀H₈O
MW: 144.17 g/mol
Boiling Pt: 75…76 °C/0.8 mm
Density: 0.990
Flash Pt: 95 °C(203 °F)
Storage Temperature: Ambient
MDL Number: MFCD00008776
CAS Number: 1817-57-8
EINECS: 217-327-1
Order Now

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty