Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

3,4-Dihydro-2H-1,5-benzodioxepin-7-amine 98%

Supplier: Thermo Scientific Chemicals
Warning
Synonyms: 7-Amino-3,4-dihydro-2H-1,5-benzodioxepine

H27862-06 H27862-03
AAH27862-06EA 212.99 USD InStock
AAH27862-06 AAH27862-03
3,4-Dihydro-2H-1,5-benzodioxepin-7-amine 98%
3,4-Dihydro-2H-1,5-benzodioxepin-7-amine


Formula: C₉H₁₁NO₂
MW: 165.19 g/mol
Boiling Pt: 123…128 °C/0.2 mm
Melting Pt: 79…81 °C
Flash Pt: >110 °C(230 °F)
Storage Temperature: Ambient
MDL Number: MFCD00218806
CAS Number: 175136-34-2
Order Now

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty