Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Trimethoxy(pentamethylcyclopentadienyl)titanium(IV) ≥97%

Supplier: Thermo Scientific Chemicals
Warning
Synonyms: Pentamethylcyclopentadienyltitanium trimethoxide

H27885-03 H27885-06
AAH27885-03EA 127.99 USD InStock
AAH27885-03 AAH27885-06
Trimethoxy(pentamethylcyclopentadienyl)titanium(IV) ≥97%
Trimethoxy(pentamethylcyclopentadienyl)titanium(IV)


Formula: C₁₃H₂₄O₃Ti
MW: 276.199 g/mol
Boiling Pt: 285 °C
Density: 1.081
Flash Pt: 61 °C(142 °F)
Storage Temperature: Ambient
MDL Number: MFCD00269850
CAS Number: 123927-75-3
UN: 1993
ADR: "3",III
Order Now

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty