Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

1-tert-Butyl-3-methylpiperazine-1,3-dicarboxylate

Supplier: Thermo Scientific Chemicals
Warning
Synonyms: 1-tert-Butyl 3-methyl piperazine-1,3-dicarboxylate

H27886-03 H27886-06
AAH27886-03EA 166.99 USD InStock
AAH27886-03 AAH27886-06
1-tert-Butyl-3-methylpiperazine-1,3-dicarboxylate
1-tert-Butyl-3-methylpiperazine-1,3-dicarboxylate


Formula: C11H20N2O4
MW: 244.29 g/mol
Storage Temperature: Ambient
MDL Number: MFCD01632465
CAS Number: 129799-08-2
Order Now

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty