Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

5-Aminopyridin-2(1H)-one 97%

Supplier: Thermo Scientific Chemicals
Warning

H27961-06 H27961-03
AAH27961-03EA 89.09 USD InStock
AAH27961-03 AAH27961-06
5-Aminopyridin-2(1H)-one 97%
5-Aminopyridin-2(1H)-one


Formula: C₅H₆N₂O
Melting Pt: ca 180 °C (dec.)
Storage Temperature: Ambient
MDL Number: MFCD03427652
CAS Number: 33630-94-3
Order Now

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty