Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

2-Cyclohexylethanamine 97%

Supplier: Thermo Scientific Chemicals
Danger
Synonyms: (2-Aminoethyl)cyclohexane

H27987-03 H27987-06
AAH27987-06EA 435.99 USD InStock
AAH27987-06 AAH27987-03
2-Cyclohexylethanamine 97%
2-Cyclohexylethanamine


Formula: C₈H₁₇N
MW: 127.23 g/mol
Storage Temperature: Ambient
MDL Number: MFCD01734731
CAS Number: 4442-85-7
EINECS: 224-673-7
UN: 2735
ADR: "8",III
Order Now

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty